Mouse Recombinant Macrophage migration inhibitory factor
Supplier: Bon Opus Biosciences
CH27-10UG CH27-1MG CH27-500UG CH27-50UG
102499-558EA 173.8 CAD
102499-558 102499-560 102509-684 102509-686
Mouse Recombinant Macrophage migration inhibitory factor
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Macrophage migration inhibitory factor is produced by our E.coli expression system and the target gene encoding Pro2-Ala115 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.