Mouse Recombinant Interleukin-13
Supplier: Bon Opus Biosciences
CH18-10UG CH18-1MG CH18-500UG CH18-50UG
102509-166EA 249.64 CAD
102509-166 102509-168 102509-680 102509-682
Mouse Recombinant Interleukin-13
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Interleukin-13 is produced by our E.coli expression system and the target gene encoding Ser26-Phe131 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.