Human Recombinant Methylated-DNA--Protein-Cysteine Methyltransferase
Supplier: Bon Opus Biosciences
CG86-10UG CG86-1MG CG86-500UG CG86-50UG
102499-352EA 158 CAD
102499-352 102499-354 102499-356 102509-536
Human Recombinant Methylated-DNA--Protein-Cysteine Methyltransferase
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human MGMT is produced by our E.coli expression system and the target gene encoding Met1-Asn207 is expressed with a 6His tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.