Human Recombinant ATP-binding cassette sub-family B member 5
Supplier: Bon Opus Biosciences
CG47-10UG CG47-1MG CG47-500UG CG47-50UG
102499-124EA 249.64 CAD
102499-124 102499-126 102499-128 102499-130
Human Recombinant ATP-binding cassette sub-family B member 5
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human ATP-binding cassette sub-family B member 5 is produced by our E.coli expression system and the target gene encoding Ile141-Val247 is expressed with a Trx tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.