Mouse Recombinant CLA
Supplier: Bon Opus Biosciences
CX92-10UG CX92-1MG CX92-500UG CX92-50UG
102498-242EA 158 CAD
102498-242 102498-244 102498-246 102498-248
Mouse Recombinant CLA
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse P-selectin glycoprotein ligand 1 is produced by our Mammalian expression system and the target gene encoding Gln42-Cys307 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.