Human Recombinant Receptor tyrosine-protein kinase erbB-2
Supplier: Bon Opus Biosciences
CX68-10UG CX68-1MG CX68-500UG CX68-50UG
102509-010EA 158 CAD
102509-010 102509-012 102509-014 102509-016
Human Recombinant Receptor tyrosine-protein kinase erbB-2
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human ErbB2 is produced by our Mammalian expression system and the target gene encoding Thr23-Thr652 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.