Human Recombinant CD27 antigen is also known as CD27L Receptor
Supplier: Bon Opus Biosciences
CX60-10UG CX60-500UG CX60-50UG
102498-110EA 66.36 CAD
102498-110 102498-114 102498-116
Human Recombinant CD27 antigen is also known as CD27L Receptor
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human CD27 is produced by our Mammalian expression system and the target gene encoding Thr21-Ile192 is expressed with a Fc, 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.