Human Recombinant Tumor Necrosis Factor Receptor Superfamily Member 3
Supplier: Bon Opus Biosciences
CX38-10UG CX38-1MG CX38-500UG CX38-50UG
102497-976EA 74.26 CAD
102497-976 102497-978 102497-980 102497-982
Human Recombinant Tumor Necrosis Factor Receptor Superfamily Member 3
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Lymphotoxin beta receptor is produced by our Mammalian expression system and the target gene encoding Gln31-Met227 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.