Human Recombinant Interleukin-10
Supplier: Bon Opus Biosciences
CX04-10UG CX04-1MG CX04-500UG CX04-50UG
102497-780EA 249.64 CAD
102497-780 102497-782 102497-784 102497-786
Human Recombinant Interleukin-10
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Interleukin-10 is produced by our Mammalian expression system and the target gene encoding Ser19-Asn178 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.