Human Recombinant Interleukin-10 Receptor Subunit beta(IL10RB)
Supplier: Bon Opus Biosciences
CC42-10UG CC42-1MG CC42-500UG CC42-50UG
102497-438EA 83.74 CAD
102497-438 102497-440 102497-442 102497-444
Human Recombinant Interleukin-10 Receptor Subunit beta(IL10RB)
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human IL-10 receptor beta is produced by our Mammalian expression system and the target gene encoding Met20-Ser220 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.