Recombinant COLEC7collectin-7
Supplier: Bon Opus Biosciences
CB54-10UG CB54-1MG CB54-50UG CB54-500UG
103529-232EA 124.82 CAD
103529-232 103529-358 103529-442 103529-484
Recombinant COLEC7collectin-7
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Pulmonary Surfactant-associated Protein D is produced by our Mammalian expression system and the target gene encoding Ala20-Phe374 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.