Recombinant Adult folate-binding protein
Supplier: Bon Opus Biosciences
CB52-10UG CB52-1MG CB52-50UG CB52-500UG
103529-230EA 124.82 CAD
103529-230 103529-356 103529-440 103529-482
Recombinant Adult folate-binding protein
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Folate Receptor Alpha is produced by our Mammalian expression system and the target gene encoding Thr25-Ser232 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.