Recombinant Human Poliovirus Receptor, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
CA99-10UG CA99-50UG CA99-500UG CA99-1MG
102943-642EA 158 CAD
102943-642 102943-644 102943-646 102943-648
Recombinant Human Poliovirus Receptor, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Poliovirus Receptor is produced by our Mammalian expression system and the target gene encoding Ala23-Leu131 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.