Human Recombinant Interleukin-17F
Supplier: Bon Opus Biosciences
CA22-10UG CA22-50UG CA22-500UG CA22-1MG
102943-458EA 173.8 CAD
102943-458 102943-460 102943-462 102943-464
Human Recombinant Interleukin-17F
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Interleukin-17F is produced by our Mammalian expression system and the target gene encoding Arg31-Gln163 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.