Recombinant Human Porphobilinogen Deaminase, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
C945-10UG C945-50UG C945-500UG C945-1MG
102943-338EA 249.64 CAD
102943-338 102943-340 102943-342 102943-344
Recombinant Human Porphobilinogen Deaminase, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Porphobilinogen Deaminase is produced by our Mammalian expression system and the target gene encoding Ser2-His361 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.