Human Recombinant Trans-2-Enoyl-CoA Reductase
Supplier: Bon Opus Biosciences
C917-10UG C917-50UG C917-500UG C917-1MG
102943-242EA 249.64 CAD
102943-242 102943-244 102943-246 102943-248
Human Recombinant Trans-2-Enoyl-CoA Reductase
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Trans-2-Enoyl-CoA Reductase is produced by our Mammalian expression system and the target gene encoding Pro54-Met373 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Order Now
SPECIFICATIONS
Enzyme name Trans-2-Enoyl-CoA Reductase
Environmentally Preferable
Enzyme type Recombinant
Source
Species Human