Human Recombinant Netrin-G1
Supplier: Bon Opus Biosciences
C916-10UG C916-50UG C916-500UG C916-1MG
102943-234EA 173.8 CAD
102943-234 102943-236 102943-238 102943-240
Human Recombinant Netrin-G1
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Netrin-G1 is produced by our Mammalian expression system and the target gene encoding His29-Ser409 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Order Now
SPECIFICATIONS
Enzyme name Netrin-G1
Environmentally Preferable
Enzyme type Recombinant
Source
Species Human