Recombinant Human Lactotransferrin/LTF
Supplier: Bon Opus Biosciences
C900-10UG C900-50UG C900-500UG C900-1MG
102943-210EA 173.8 CAD
102943-210 102943-212 102943-214 102943-216
Recombinant Human Lactotransferrin/LTF
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant Human Lactotransferrin/LTF is produced by our Mammalian expression system and the target gene encoding Gly20-Lys710 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx.
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.