Mouse Recombinant Kit ligand
Supplier: Bon Opus Biosciences
C775-10UG C775-50UG C775-500UG C775-1MG
102942-842EA 186.44 CAD
102942-842 102942-844 102942-846 102942-848
Mouse Recombinant Kit ligand
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Stem Cell Factor is produced by our E.coli expression system and the target gene encoding Lys26-Ala189 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.