Human Recombinant Teratocarcinoma-derived Growth Factor 1
Supplier: Bon Opus Biosciences
C762-10UG C762-50UG C762-500UG C762-1MG
102942-738EA 249.64 CAD
102942-738 102942-740 102942-742 102942-744
Human Recombinant Teratocarcinoma-derived Growth Factor 1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Teratocarcinoma-derived growth factor 1 is produced by our Mammalian expression system and the target gene encoding Leu31-Ser169 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.