Human Recombinant Serine protease HTRA2

Supplier: Bon Opus Biosciences

C760-10UG C760-50UG C760-500UG C760-1MG
102942-722EA 83.74 CAD
102942-722 102942-724 102942-726 102942-728
Human Recombinant Serine protease HTRA2
Proteins and Peptides

Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products


  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels


Recombinant Human HTRA2 is produced by our Mammalian expression system and the target gene encoding Ala134-Glu458 is expressed with a 6His tag at the C-terminus.


Accessories information: Accessories not included.


Delivery information: Shipped Via FedEx


Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.


Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.

The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.

Order Now

SPECIFICATIONS

Protein/peptide name Serine protease HTRA2
Environmentally Preferable
Protein/peptide type Recombinant
Species Human
Source

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR