Recombinant Human Neuregulin-1 beta, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
C753-10UG C753-50UG C753-500UG C753-1MG
102942-434EA 158 CAD
102942-434 102942-436 102942-438 102942-440
Recombinant Human Neuregulin-1 beta, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Neuregulin-1 beta is produced by our E.coli expression system and the target gene encoding Ser2-Lys246 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.