Mouse Tff1 (C-6His)-, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
C660-10UG C660-50UG C660-500UG C660-1MG
103529-236EA 150.1 CAD
103529-236 103529-270 103529-344 103529-546
Mouse Tff1 (C-6His)-, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Trefoil Factor 1 is produced by our Mammalian expression system and the target gene encoding Gln22-Phe87 is expressed with a 6His at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.