Lab Stand Support Shelf, Ace Glass
Supplier: Ace Glass
11173-04 11173-06
76214-470EA 774.46 CAD
76214-470 76214-472
Lab Stand Support Shelf, Ace Glass
Supports Flask Supports
Lab stand support shelf fits any stand with up to 5/8" O.D. rod. Rod-mounted, and features a 5/8" O.D. × 12" long stainless rod with a through hole.
  • Manufactured from light, cast aluminum and epoxy coated
Small platform is for use with up to #50 Ace-Thred adapters. Large platform is for use with up to #80 Ace-Thred adapters.
Order Now
SPECIFICATIONS
Environmentally Preferable