Densito and DensitoPro Handheld Density Meters, METTLER TOLEDO®
Supplier: Mettler Toledo
30330857 30330858
76213-948EA 6087.35 CAD
76213-948 76213-950
Densito and DensitoPro Handheld Density Meters, METTLER TOLEDO®
Density Meters
Designed for use in the lab and on the go, the Densito and DensitoPro are perfect for all applications.
  • Perfect for quality control of incoming goods to at-line quality assurance of intermediate or final products
  • Users can be guided through multi-step procedures with clear directions, from measurement to cleaning
Accessories information: A wide assortment of accessories are available, but must be ordered separately. These include a standard filling tube, as well as a connector syringe/filling tube for use with viscous samples. A carrying case, USB-C charging cable, and power supply with worldwide adapters are also available.