MycoZap™ Plus-PR
Supplier: LONZA PHARMA - BIOSCIENCE CA
VZA-2022 VZA-2021 VZA-2021 VZA-2022
CA10064-508EA 689.28 CAD
CA10064-508 75870-440 75870-442 CA10064-506
MycoZap™ Plus-PR
MycoZap™ Plus-PR
MycoZap™ Antibiotics are extremely powerful combinations of innovative antibiotics for the protection of cell cultures from mycoplasma contamination. While MycoZap™ Prophylactic prevents mycoplasma contamination, MycoZap™ Plus offers complete protection against a broad range of common contaminants including mycoplasma.
  • Active against mycoplasma, Gram(-) and Gram(+) bacteria as well as yeast and fungi.
  • Complete solution replacing Pen/Strep formulation.
Applications: MycoZap™ Plus-PR optimized for gentle protection of primary cells.