MycoZap™ Plus-CL
Supplier: LONZA PHARMA - BIOSCIENCE CA
VZA-2011 VZA-2012 VZA-2012 VZA-2011
CA10064-594EA 385.13 CAD
CA10064-594 CA10064-504 75870-438 75870-448
MycoZap™ Plus-CL
MycoZap™ Plus-CL
MycoZap™ Antibiotics are extremely powerful combinations of innovative antibiotics for the protection of cell cultures from mycoplasma contamination. While MycoZap™ Prophylactic prevents mycoplasma contamination, MycoZap™ Plus offers complete protection against a broad range of common contaminants including mycoplasma.
Benefits:
Active against mycoplasma, Gram(-) and Gram(+) bacteria as well as yeast and fungi
Complete solution replacing Pen/Strep formulation
Applications:
MycoZap™ Plus-CL for protection of cell lines