Amphotericin B 250 µg/mL in aqueous solution, HyClone™

Supplier: Cytiva

Synonyms: (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid

SV30078.01 SV30078.01
CA82026-728EA 78.17 CAD
CA82026-728 82026-728
Amphotericin B 250 µg/mL in aqueous solution, HyClone™
Amphotericin B

An antifungal, anti-yeast and anti-mold reagent.


  • An antibiotic agent that is active against fungi
  • Interferes with the permeability of the cell membrane of sensitive fungi by binding sterols
  • Common working concentration is 2.5 μg/mL, and it is cytotoxic at 30 μg/mL

Formula: C₄₇H₇₃NO₁₇
MW: 924.08 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00877763
CAS Number: 1397-89-3

Order Now

Specification Test Results

Sterilization 0.2 µm sterile filtered
Antibiotic Spectrum Antimycotic, Antifungal
Concentration 250 µg/mL

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR