Beakers, Jacketed, Conical Bottom, Chemglass
Supplier: Chemglass
CG-1103-05 CG-1103-06 CG-1103-01 CG-1103-02 CG-1103-03 CG-1103-04
80066-224EA 1027.9 CAD
80066-224 80066-226 80066-216 80066-218 80066-220 80066-222
Beakers, Jacketed, Conical Bottom, Chemglass
Beakers
Designed for use in temperature controlled chemical and biological reactions.
Bottom interior surface is conical in shape to help eliminate spinout when used with egg shaped spinbars. Hose connections have an O.D. of 10 mm at the largest serration.