Mini Economy Splash Goggles
Supplier: DR INSTRUMENTS INC. SE
DR83210 DR83010 DR83000 DR83200
470303-202CS 792.4 CAD
470303-202 470303-204 470303-198 470303-200
Mini Economy Splash Goggles
Goggles
Perfect for elementary students with small faces.
  • Direct and Indirect vent styles available
  • Anti-fog coating available
  • UV absorbing polycarbonate lens
Lightweight, green tinted PVC body gently conforms to the face, providing a secure and comfortable fit. Ideal for smaller faces and working in confined areas. Direct vent provides maximum air flow and protection against grinding, chipping, and cutting operations.