Top Dog 8 Mil Die Cut Apron, 50" Length, Remco Products

Supplier: Remco Products
81026 81022 81023 81024 81025
76196-524CS 1250.65 CAD
76196-524 76196-582 76196-584 76196-586 76196-588
Top Dog 8 Mil Die Cut Apron, 50" Length, Remco Products
Workwear
Reusable and washable, the one-piece construction increases the apron's hygiene and durability.

  • Resistant to chemical and acid sanitation products
  • Non-porous surface will not host bacteria or mold
  • Washer and dryer safe
  • Will not crack or stiffen in low temperatures or with repeated cleaning
  • Impervious to animal fats and degreasers
  • Completely latex-free for sensitive users
  • Easily repairable

8 mil aprons from Top Dog come in five colors to support color-coded HACCP plans and zoning efforts. The 8 Mil Die Cut Apron is made of pure polyurethane and is reversible. It fits comfortably over clothes. The apron is machine washable, dryer-safe to 180 °F, and chemical and abrasive resistant. The 50" length makes the apron cover most of an employee's torso and legs without becoming cumbersome.

Certifications: Made of FDA Compliant material.

Ordering information: Aprons can only be purchased by the case. Case quantity is 24.

Delivery information: Dropshipping available.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR