Anti-IgG Rabbit Polyclonal Antibody

Supplier: Immunoreagents
IR2387
10151-628EA 297.37 CAD
10151-628
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
  • Origin: US

Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR