3,5-Di-tert-butyl-4-hydroxycinnamic acid predominantly trans 98%

Supplier: Thermo Scientific Chemicals
Warning

A10514-09 A10514-18
CAAAA10514-09EA 120.08 CAD
CAAAA10514-09 CAAAA10514-18
3,5-Di-tert-butyl-4-hydroxycinnamic acid predominantly trans 98%
3,5-Di-tert-butyl-4-hydroxycinnamic acid
Formula: C₁₇H₂₄O₃
Melting Pt: ∼ 195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00017291
CAS Number: 22014-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR