Polyethylene glycol monomethyl ether, M.W. 550 550

Supplier: TCI America

P2184-100G P2184-500G
TCP2184-100GEA 58.46 CAD
TCP2184-100G TCP2184-500G
Polyethylene glycol monomethyl ether, M.W. 550 550
Polyethylene glycol monomethyl ether
Formula: CH₃(OCH₂CH₂)nOH
Flash Pt: 182 °C (lit.)
MDL Number: MFCD00084416
CAS Number: 9004-74-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55d77bce8ce849bacaf24aa2d908b158985d327a&vid=55d77bce8ce849bacaf24aa2d908b158985d327a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Average molecular weight 525 to 575
Hydroxyl value 95 to 107


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR