4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

C2816-1G C2816-200MG
TCC2816-1GEA 145.52 CAD
TCC2816-1G TCC2816-200MG
4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide ≥98.0% (by HPLC, titration analysis)
4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide
Formula: C₁₇H₁₄F₃N₃O₂S
MDL Number: MFCD00941298
CAS Number: 169590-42-5
Merck Index: 14,01956
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cf9f87d30d9e532b73258a0e6eb6df0cb549518&vid=1cf9f87d30d9e532b73258a0e6eb6df0cb549518&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (HPLC) Min. 98.0 area%
Purity(Neutralization titration) Min. 98.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR