Airfree® Reaction Flasks, Round Bottom, Schlenk, Chemglass
Supplier: Chemglass
AF-0528-07 AF-0528-26 AF-0528-01 AF-0528-02 AF-0528-03 AF-0528-04 AF-0528-05 AF-0528-06 AF-0528-20 AF-0528-21 AF-0528-22 AF-0528-23 AF-0528-24 AF-0528-25
80064-660EA 454.16 CAD
80064-660 80064-674 80064-648 80064-650 80064-652 80064-654 80064-656 80064-658 80064-662 80064-664 80064-666 80064-668 80064-670 80064-672
Airfree® Reaction Flasks, Round Bottom, Schlenk, Chemglass
Reaction Vessels Reaction Vessels, Flask Shape
These round bottom single neck flask having a standard taper outer joint.
Available with a choice of a 2 mm high vacuum glass stopcock having a S.S. constant pressure retaining device or a 0 to 4 mm Chem-Cap® Airfree® inlet valve.