ClikLok™ and Screw Cap MacrewTube™ Microcentrifuge Tube Accessories, 5.0 ml, Simport Scientific
Supplier: Simport Scientific
T330-25B T330-25G T330-25P M957B M957G M957L M957R M957Y T330-75CLB T330-75CLG T330-75CLR T330-75CLW T330-75CLY T330-25Y T330AD15 T330AD50 T450-25B T450-25O T450-25R
76209-766CS 294.94 CAD
76209-766 76209-768 76209-770 76209-828 76209-830 76209-832 76209-834 76209-836 76209-838 76209-840 76209-842 76209-844 76209-846 76209-848 76209-858 76209-860 76209-862 76209-864 76209-866
ClikLok™ and Screw Cap MacrewTube™ Microcentrifuge Tube Accessories, 5.0 ml, Simport Scientific
Tubes Microcentrifuge Tubes
Color-coded cap inserts, CapLock™ Clips, centrifuge adaptors, and storage racks are all available options designed for use with ClikLok™ and MacrewTube™ microcentrifuge tubes.
The color-coded CapInsert™ can be applied to the tube closure. Manually press fit and lock into place on top of closure.
The CapLock™ Clip for 5.0 ml Tubes provides extra security against inadvertent cap openings on critical samples or during incubation above 80 °C. They are ideal for shipping samples. Made of polyethylene.
The Univesal Centrifuge adapters are made of acetal plastic. Reusable and freezable (upto –90 °C), they are offered in both a 15 and 50 ml rotor bore size.
The T425 Series Rack is reusable, autoclavable, and freezable (to –90 °C). Wells are numbered for easier identification. Racks will accommodate 25 tubes.
The 5.0 ml Tube Storage Box allows samples to be stored from –90 to 80 °C. A transparent cover allows users to see the contents of the box, and is keyed to the base to prevent misalignment. Printed with a series of squares, numbered 1 to 25, the surface accepts writing with markers to faciliatate inventory control. A unique color-coding system uses colored plastic grids to separate the cover from the base.