pH Buffer Solutions, NIST
Supplier: METTLER TOLEDO(BALANCES) CA
30258823
30258827
30258811
10804-850EA
46.87
CAD
10804-850
10804-858
10804-868
pH Buffer Solutions, NIST
Buffers
pH Calibration Buffers
These buffer solutions are used for calibration and verification of measurement system.
- Shelf life: 24 months
- Uncertainty: ±0.02 pH
Certifications: A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...