ClikLok™ and Screw Cap MacrewTube™ Microcentrifuge Tubes, Simport Scientific
Supplier: Simport Scientific
T330-75NS T330-75AMS T330-76N T330-76B T330-76G T330-76R T330-76Y T330-76AM T330-75Y T330-75AM T330-76NS T330-76AMS T330-75N T330-75B T330-75G T330-75R T366NLS T366BLS T366GLS T366RLS T366WLS T366YLS T366AMLS T366NLSL T366BLSL T366GLSL T366RLSL T366WLSL T366YLSL T366AMLSL T365-5TN T365-5TAM T365-5AMLSS
76209-750CS 118.66 CAD
76209-750 76209-752 76209-754 76209-756 76209-758 76209-760 76209-762 76209-764 76209-784 76209-786 76209-788 76209-790 76209-792 76209-794 76209-796 76209-798 76209-800 76209-802 76209-804 76209-806 76209-808 76209-810 76209-812 76209-814 76209-816 76209-818 76209-820 76209-822 76209-824 76209-826 76209-850 76209-852 76209-854 76209-856
ClikLok™ and Screw Cap MacrewTube™ Microcentrifuge Tubes, Simport Scientific
Tubes Microcentrifuge Tubes
The 5.0 ml ClikLok™ and MacrewTube™ microcentrifuge tubes have been designed for the simple and safe processing of medium-sized sample volumes.
  • Made of high-quality, transparent polypropylene without use of slip agents, plasticizers, mold release agents, and biocide substances
  • Centrifuge up to 25000 ×g
  • RNase, DNase, Pyrogen, and DNA-free
  • Operating temperature range from –196 to 121 °C
  • Autoclavable
  • Use the color-coded CapInsert™ for easy identification
Designed for samples larger than 2.0 ml. The 16mm tube diameter is identical to the diameter of standard 15 ml tubes, ensuring further compatibility with centrifuge rotors. Both the ClikLok™ and MacrewTube™ are designed with a smooth bottom with no sharp points to prevent hurting user fingers. The slim design fits high capacity rotors. Tubes are graduated every 0.5 ml.
The ClikLok™ snap cap tube is available with a flat top or with a CapInsert™ cavity for color-coding purposes. The cap will partially open when the internal pressure approaches an unsafe level in a controlled manner, releasing excess pressure but preventing any splashing of contents. In applications over 80 °C, the CapLock™ Clips are recommended.
The screw cap MacrewTube™ is available sterile and with a choice of two caps, either a regular lip seal cap or a tethered cap. Both models have the CapInsert™ cavity for color-coding purposes.
Ordering information: Color coding CapInserts™, Caplock™ Clips, universal adapters, racks, and storages boxes are available seperately. See the 5.0 ml ClikLok™ and Screw Cap MacrewTube™ Microcentrifuge Tube Accessories page for details.
Packaging: Sterile products are packaged in bags of 20. Nonsterile tubes are packaged in bags of 100.