Nonidet® P 40 Substitute (NP-40) 10% aqueous solution, 2D-Detergent™

Supplier: G-Biosciences
Warning

DG901 DG902
CA95057-618EA 603.78 CAD
CA95057-618 CA95057-620
Nonidet® P 40 Substitute (NP-40) 10% aqueous solution, 2D-Detergent™
Nonidet® P 40 Substitute (NP-40)

Many commercial grade detergents contain elevated levels of sulfhydryl oxidizing agents, peroxides, salts and carbonyl compounds


  • Low conductivity; <15µS
  • Low peroxide contamination.
  • Low carbonyl contamination.
  • Reduced metal ions.
  • Sterile detergent solution
  • 10% aqueous solutions.
  • Sealed under inert gas to prevent oxidation.


2D-Detergent™ solutions contain reduced peroxides and carbonyl compounds. In addition, the detergents have less than 15µS conductivity. These detergents are offered as 10% aqueous solutions, sealed under inert gas and are suitable for all protein applications, including 2D-electrophoresis. These non-ionic detergents are suitable for isolating membrane-protein complexes.


The proteins that are isolated with these detergents are highly susceptible to contaminating peroxides and carbonyls. The peroxides will oxidize proteins and the carbonyl groups will form Schiff's bases with the proteins that interfere with a protein's structure.


The aldehyde and peroxide levels are <100µM and <10µM respectively with a conductivity of <15µS.


Ordering information: Accessories not included.

Storage Temperature: Ambient
MDL Number: MFCD00132851
CAS Number: 9016-45-9
EINECS: 500-024-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR