Minerals Model Set
Supplier: Mega Molecules
GC430
470232-074EA 97.2 CAD
470232-074
Minerals Model Set
Models Crystal Models
Hands on mineral crystal structures.
  • Includes 1 activity
  • Perfect for high school and college students
Build the most abundant silicate minerals including feldspar, quartz, olivine, pyroxenes, amphiboles, micas, and clay minerals. Learn how models of silicate minerals are related to unit cell and crystal lattice structures. This model set has enough parts to construct all six types of silicate minerals at one time allowing students to compare nesosilicates, sorosilicates, cyclosilicates, inosilicates, phyllosilicates, and tectosilicates.