ALPS™ Heat Sealer, Thermo Scientific
Supplier: Thermo Scientific
ALPS™
AB-1443A
89499-572CS 11093.88 CAD
89499-572
ALPS™ Heat Sealer, Thermo Scientific
Heat Sealers Bag Sealers
ALPS™ Microplate heat sealer is designed to provide sealing consistency in low to medium throughput labs.
  • Offers superior sample protection of storage and reaction plates
  • Applications includes compound storage, sample archiving and PCR
  • Simple on-board touch pad controls adjust sealing temperature and time for reproducible results
  • Audible alarm indicates sealing completion
  • Lighweight: 7.2 kg
Apply consistent, secure, tight seals around individual wells, eliminating sample loss through evaporation and cross contamination between wells.
Counter lever handle design provides minimal strain when operating the instrument.