Chloroquine diphosphate salt ≥99% (by titrimetric analysis), crystalline powder BP
Supplier: CHEM-IMPEX INTERNATIONAL, INC
Synonyms:
N4-(7-Chloroquinolin-4-yl)-N1,N1-diethylpentane-1,4-diamine bis(phosphate), N4-(7-Chloro-4-quinolyl)-N1,N1-diethyl-1,4-pentanediamine Diphosphate, 7-Chloro-4-[4-(diethylamino)-1-methylbutylamino]quinoline Diphosphate, Chloroquine phosphate
22113-250G
22113-5G
CH22113-250GEA
544.66
CAD
CH22113-250G
CH22113-5G
Chloroquine diphosphate salt ≥99% (by titrimetric analysis), crystalline powder BP
Chloroquine diphosphate salt
Formula:
C₁₈H₂₆ClN₃ MW: 515.86 g/mol Melting Pt: 193…197 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00069852 CAS Number: 50-63-5 |
Specification Test Results
Assay by titration | ≥99% |
Purity by UV | Conforms |
pH | 3.8 - 4.3 |
Infrared spectrum | Conforms to structure |
Heavy metals | ≤20 ppm |
Water content (Karl Fisher) | ≤2.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...