Twirl'Blue PE Gloves, Sterile, Labplas
Supplier: Labplas
SA7-GLOVES
75849-336CS 191.09 CAD
75849-336
Twirl'Blue PE Gloves, Sterile, Labplas
Gloves
These gamma sterilized blue color gloves are made of polyethylene and specifically designed for food industries to avoid contaminations.
  • R-Nase, D-Nase and pyrogen free
  • Accommodate solid, semi-solid and liquid compounds
  • Food contact approved
The main purpose of the distinct color design is to avoid possible foreign matter contamination and for increased visibility.