Caps for Mobile Phase Bottles, Microsolv
Supplier: MicroSolv Technology Corp
58807-B45 58807-G45 58807-P45 58807-Y45
89219-970PK 94.71 CAD
89219-970 89219-972 89219-974 89219-976
Caps for Mobile Phase Bottles, Microsolv
Closures Bottle Closures
These caps are designed for use on HPLC mobile phase reservoir and storage bottles with GL 45 thread.
  • Made up of polypropylene