Oblong HDPE Plastic Bottles, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W218810 W218811 W218812 W218813 W218814 W218815 W218816 W218817 W218818 W218819
10847-638CS 506.36 CAD
10847-638 10847-640 10847-642 10847-644 10847-646 10847-648 10847-650 10847-652 10847-654 10847-656 10847-658
Oblong HDPE Plastic Bottles, DWK Life Sciences
Bottles Plastic Bottles
HDPE Oblong bottles with or without cap attached.
  • Good chemical resistance
  • Natural in color
Wide mouth ideal for storage and management of solid samples and powders.