Rushton Impeller, R-100, Lightnin®
Supplier: Lightnin
809414 809415
58958-071EA 2622.63 CAD
58958-071 58958-073
Rushton Impeller, R-100, Lightnin®
Stirrers Stirring Tools
These radial flow impellers are designed with classic 90° flat blade turbine.
  • Recommended for high shear mixing and gas dispersion applications
  • Customizable using varying blade widths and lenghts
The unique feature of the blades provides shear to achieve good contacting for liquid-liquid and gas-liquid dispersions and emulsions.