Dippers, PTFE, Dynalon
Supplier: Dynalon
307024-0100 307024-0250 307024-1000 307024-HAND
76210-792EA 413.59 CAD
76210-792 76210-794 76210-818 76210-820
Dippers, PTFE, Dynalon
Samplers Sample Dippers
Totally inert and non-contaminating PTFE dipper sampler.
Extendable screw-in shafts have a steel core for rigidity and are fully encapsulated in PTFE.
Delivery information: The basic unit consists of a beaker with a single 24" (600 mm) shaft. Replacement shaft is available separately.