Dippers, HDPE, Dynalon
Supplier: Dynalon
107075 107095 106505 106515 106525
76210-676EA 164.16 CAD
76210-676 76210-690 76210-692 76210-694 76210-696
Dippers, HDPE, Dynalon
Samplers Sample Dippers
High density polyethylene dipper for removing samples from large tanks.
  • Graduated plastic dipper
  • 2 pour spouts
  • PVC hand grip on 1" diameter HDPE rod
  • 500 mL and 1000 mL beaker welded to the rigid handle at a 45° angle
Made in the USA, these plastic dippers are lightweight but durable.
Packaging: The 12' size is supplied with a two-piece handle for economy in shipping and storage.