Liquid Sampler, HDPE, Dynalon
Supplier: Dynalon
114505-0500 114515-0500
76210-788EA 565.94 CAD
76210-788 76210-790
Liquid Sampler, HDPE, Dynalon
Samplers Fluid Samplers
This sampling device is perfect for corrosive environments and for hard to reach places.
  • Excellent for industrial and environmental applications
Plastic Liquid Sampler is manufactured from chemically resistant high density polyethylene.
The included bottles screw into the end of the handle and is filled via four 5mm diameter ports (2 allowing liquid to enter and 2 allowing air to escape). All rods are 1.25" O.D.
Packaging: Two-piece handle for UPS shipment.
Order Now
SPECIFICATIONS
Environmentally Preferable